Assay Details
| Gene Symbol: ARL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004311.3 | NP_004302.1 | 6 | 3333 | 85 | |
| XM_017016260.1 | XP_016871749.1 | 6 | 3519 | 85 | ||
| GenBank mRNA | AF038193.1 | - | 1 | 1032 | 85 | |
More Information
Set Membership: |
|

ARL3
Genomic Map