Assay Details
| Gene Symbol: CHMP1B2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110646.1 | 6 | 3860 | 95 | ||
| GenBank mRNA | CR627394.1 | - | 6 | 3859 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHMP1B2P| Gene Symbol: CHMP1B2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110646.1 | 6 | 3860 | 95 | ||
| GenBank mRNA | CR627394.1 | - | 6 | 3859 | 95 | |
Set Membership: |
|