Assay Details
| Gene Symbol: DDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092374.1 | - | 1 | 2958 | 81 | |
| AK093865.1 | - | 1 | 1217 | 81 | ||
| BX537651.1 | - | 1 | 1327 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092374.1 | - | 1 | 2958 | 81 | |
| AK093865.1 | - | 1 | 1217 | 81 | ||
| BX537651.1 | - | 1 | 1327 | 81 | ||
Set Membership: |
|