Assay Details
| Gene Symbol: LOC105378269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134498.1 | 2 | 495 | 101 | ||
| GenBank mRNA | BM682592.1 | - | 1 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105378269| Gene Symbol: LOC105378269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134498.1 | 2 | 495 | 101 | ||
| GenBank mRNA | BM682592.1 | - | 1 | 101 | ||
Set Membership: |
|