Assay Details
| Gene Symbol: SRSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017000100.1 | XP_016855589.1 | 3 | 1353 | 157 | |
| GenBank mRNA | BC071575.1 | - | 3 | 756 | 160 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017000100.1 transcripts.
Set Membership: |
|

SRSF10
Genomic Map