Assay Details
| Gene Symbol: NTRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006720543.3 | XP_006720606.1 | 17 | 4923 | 93 | |
| XM_006720544.3 | XP_006720607.1 | 16 | 4899 | 93 | ||
| XM_017022240.1 | XP_016877729.1 | 18 | 4794 | 93 | ||
| XM_017022241.1 | XP_016877730.1 | 17 | 4279 | 93 | ||
| XM_017022243.1 | XP_016877732.1 | 17 | 4275 | 93 | ||
| XM_017022244.1 | XP_016877733.1 | 17 | 4177 | 93 | ||
| XM_017022245.1 | XP_016877734.1 | 18 | 4232 | 93 | ||
| XM_017022246.1 | XP_016877735.1 | 17 | 4333 | 93 | ||
| XM_017022251.1 | XP_016877740.1 | 10 | 3294 | 93 | ||
| XM_017022252.1 | XP_016877741.1 | 9 | 3270 | 93 | ||
| XM_017022253.1 | XP_016877742.1 | 18 | 4965 | 93 | ||
| XM_017022254.1 | XP_016877743.1 | 18 | 4998 | 93 | ||
More Information
Set Membership: |
|

NTRK3
Genomic Map