Assay Details
| Gene Symbol: SSTR5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027242.1 | 4 | 1652 | 68 | ||
| GenBank mRNA | AK056814.1 | - | 4 | 1596 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSTR5-AS1| Gene Symbol: SSTR5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027242.1 | 4 | 1652 | 68 | ||
| GenBank mRNA | AK056814.1 | - | 4 | 1596 | 68 | |
Set Membership: |
|