Assay Details
| Gene Symbol: NPIPB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011523501.2 | XP_011521803.1 | 1 | 412 | 94 | |
| Gene Symbol: NPIPB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128772.1 | - | 1 | 1081 | 94 | |
| Gene Symbol: PDXDC2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX647358.1 | - | 1 | 3213 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK128772.1, XM_011523501.2 transcripts.
Set Membership: |
|

NPIPB15
Genomic Map