Assay Details
| Gene Symbol: PDPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261816.1 | NP_001248745.1 | 12 | 5587 | 135 | |
| NM_002613.4 | NP_002604.1 | 14 | 5798 | 135 | ||
| NM_031268.5 | NP_112558.2 | 11 | 5417 | 135 | ||
| XM_011522521.2 | XP_011520823.1 | 14 | 9486 | 135 | ||
| XM_011522523.2 | XP_011520825.1 | 11 | 9110 | 135 | ||
| XM_017023271.1 | XP_016878760.1 | 13 | 9866 | 135 | ||
| XM_017023272.1 | XP_016878761.1 | 13 | 6422 | 135 | ||
| XM_017023273.1 | XP_016878762.1 | 13 | 6131 | 135 | ||
| XM_017023274.1 | XP_016878763.1 | 14 | 6269 | 135 | ||
| XM_017023275.1 | XP_016878764.1 | 10 | 9491 | 135 | ||
| XM_017023276.1 | XP_016878765.1 | 10 | 5735 | 135 | ||
| XM_017023277.1 | XP_016878766.1 | 11 | 5833 | 135 | ||
| GenBank mRNA | AK095507.1 | - | 1 | 1182 | 135 | |
More Information
Set Membership: |
|

PDPK1
Genomic Map