Assay Details
| Gene Symbol: CAMTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171166.1 | NP_001164637.1 | 21 | 3724 | 90 | |
| NM_001171167.1 | NP_001164638.1 | 23 | 4182 | 90 | ||
| NM_001171168.1 | NP_001164639.1 | 22 | 4112 | 90 | ||
| NM_015099.3 | NP_055914.2 | 23 | 3840 | 90 | ||
| XM_006721478.3 | XP_006721541.1 | 23 | 4487 | 90 | ||
| XM_006721481.3 | XP_006721544.1 | 22 | 4400 | 90 | ||
| XM_006721482.3 | XP_006721545.1 | 21 | 3533 | 90 | ||
| XM_011523746.2 | XP_011522048.1 | 23 | 3847 | 90 | ||
| XM_011523747.2 | XP_011522049.1 | 23 | 3831 | 90 | ||
| XM_011523748.2 | XP_011522050.1 | 22 | 3825 | 90 | ||
| XM_011523749.2 | XP_011522051.1 | 22 | 3809 | 90 | ||
| GenBank mRNA | AB020716.1 | - | 23 | 3801 | 90 | |
| AL831849.1 | - | 22 | 4112 | 90 | ||
| AL833974.1 | - | 21 | 3709 | 90 | ||
| BC010050.2 | - | 14 | 2232 | 90 | ||
| BC016163.1 | - | 10 | 1459 | 90 | ||
| BC032381.1 | - | 7 | 855 | 90 | ||
| BC136534.1 | - | 22 | 3769 | 9020524 | 90 | |
| BC142695.1 | - | 23 | 3831 | 40148717 | 90 | |
| BC144232.1 | - | 21 | 3697 | 9052752 | 90 | |
| BC167148.1 | - | 23 | 3801 | 8862515 | 90 | |
| BC167149.1 | - | 23 | 3801 | 8862516 | 90 | |
| BC167151.1 | - | 23 | 3801 | 8862527 | 90 | |
| BC167160.1 | - | 23 | 3801 | 8862594 | 90 | |
More Information
Set Membership: |
|

CAMTA2
Genomic Map