Assay Details
| Gene Symbol: CAMTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006721478.3 | XP_006721541.1 | 1 | 170 | 77 | |
| XM_006721481.3 | XP_006721544.1 | 1 | 170 | 77 | ||
| Gene Symbol: LOC101927979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123098.1 | - | 1 | 77 | ||
| DA740680.1 | - | 1 | 77 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006721478.3, XM_006721481.3 transcripts.
Set Membership: |
|

CAMTA2
Genomic Map