Assay Details
| Gene Symbol: LOC105369205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011524131.2 | XP_011522433.1 | 4 | 4492 | 114 | |
| Gene Symbol: USP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015276.1 | NP_056091.1 | 13 | 114 | ||
| XM_005256575.2 | XP_005256632.1 | 13 | 114 | |||
| GenBank mRNA | AB028986.1 | - | 13 | 114 | ||
| AK025331.1 | - | 1 | 114 | |||
| AK130677.1 | - | 1 | 114 | |||
| AL049258.1 | - | 1 | 114 | |||
| AL110213.1 | - | 1 | 114 | |||
| BC007196.1 | - | 6 | 114 | |||
| BC013364.2 | - | 1 | 114 | |||
| BX640815.1 | - | 12 | 114 | |||
More Information
Set Membership: |
|

LOC105369205
Genomic Map