Assay Details
| Gene Symbol: NSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042549.1 | NP_001036014.1 | 7 | 7446 | 76 | |
| NM_001297736.1 | NP_001284665.1 | 4 | 7238 | 76 | ||
| NM_001297737.1 | NP_001284666.1 | 5 | 7293 | 76 | ||
| NM_015471.3 | NP_056286.3 | 6 | 7361 | 76 | ||
More Information
Set Membership: |
|

NSL1
Genomic Map