Assay Details
| Gene Symbol: SPSB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025106.3 | NP_079382.2 | 3 | 1758 | 137 | |
| GenBank mRNA | AK130055.1 | - | 1 | 625 | 137 | |
| BC005852.2 | - | 2 | 922 | 137 | ||
| BC015711.1 | - | 3 | 1709 | 3916157 | 137 | |
More Information
Set Membership: |
|

SPSB1
Genomic Map