Assay Details
| Gene Symbol: LOC100131170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096606.1 | - | 1 | 1104 | 84 | |
| AK098403.1 | - | 3 | 2442 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100131170| Gene Symbol: LOC100131170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096606.1 | - | 1 | 1104 | 84 | |
| AK098403.1 | - | 3 | 2442 | 84 | ||
Set Membership: |
|