Assay Details
| Gene Symbol: ZNF519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017025562.1 | XP_016881051.1 | 6 | 14616 | 64 | |
| XM_017025563.1 | XP_016881052.1 | 7 | 10616 | 64 | ||
| XM_017025564.1 | XP_016881053.1 | 6 | 10355 | 64 | ||
| XM_017025565.1 | XP_016881054.1 | 7 | 10433 | 64 | ||
More Information
Set Membership: |
|

ZNF519
Genomic Map