Assay Details
| Gene Symbol: CEBPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285829.1 | NP_001272758.1 | 1 | 2514 | 97 | |
| NM_001287424.1 | NP_001274353.1 | 1 | 2514 | 97 | ||
| NM_001287435.1 | NP_001274364.1 | 1 | 2514 | 97 | ||
| NM_004364.4 | NP_004355.2 | 1 | 2514 | 97 | ||
| GenBank mRNA | BC063874.1 | - | 2 | 2042 | 97 | |
| X87248.1 | - | 1 | 2375 | 97 | ||
More Information
Set Membership: |
|

CEBPA
Genomic Map