Assay Details
| Gene Symbol: B3GNT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198540.2 | NP_940942.1 | 3 | 1756 | 111 | |
| XM_011526934.2 | XP_011525236.1 | 2 | 3075 | 111 | ||
| GenBank mRNA | AY277592.2 | - | 3 | 1756 | 111 | |
More Information
Set Membership: |
|

B3GNT8
Genomic Map