Assay Details
| Gene Symbol: CASP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012114.2 | NP_036246.1 | 7 | 1236 | 88 | |
| XM_011527861.1 | XP_011526163.1 | 7 | 1310 | 88 | ||
| GenBank mRNA | EU502748.1 | - | 7 | 1332 | 88 | |
More Information
Set Membership: |
|

CASP14
Genomic Map