Assay Details
| Gene Symbol: ZNF570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300993.2 | NP_001287922.1 | 6 | 3383 | 87 | |
| NM_001321991.1 | NP_001308920.1 | 5 | 3232 | 87 | ||
| NM_001321992.1 | NP_001308921.1 | 4 | 3563 | 87 | ||
| NM_001321993.1 | NP_001308922.1 | 5 | 3706 | 87 | ||
| NM_001321994.1 | NP_001308923.1 | 4 | 3622 | 87 | ||
| NM_144694.3 | NP_653295.1 | 5 | 3647 | 87 | ||
More Information
Set Membership: |
|

ZNF570
Genomic Map