Assay Details
| Gene Symbol: PPP5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB209615.1 | - | 1 | 1544 | 80 | |
| AK094241.1 | - | 1 | 273 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP5C| Gene Symbol: PPP5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB209615.1 | - | 1 | 1544 | 80 | |
| AK094241.1 | - | 1 | 273 | 80 | ||
Set Membership: |
|