Assay Details
| Gene Symbol: MYH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077186.1 | NP_001070654.1 | 42 | 6686 | 96 | |
| NM_001145809.1 | NP_001139281.1 | 43 | 6785 | 96 | ||
| NM_024729.3 | NP_079005.3 | 41 | 6662 | 96 | ||
| XM_006723386.3 | XP_006723449.1 | 42 | 6747 | 96 | ||
| XM_011527320.2 | XP_011525622.1 | 42 | 6764 | 96 | ||
| XM_011527321.2 | XP_011525623.1 | 41 | 6740 | 96 | ||
| XM_011527323.2 | XP_011525625.1 | 42 | 6754 | 96 | ||
| GenBank mRNA | AB111886.1 | - | 32 | 5344 | 96 | |
| AB290169.1 | - | 43 | 6755 | 96 | ||
| AK023943.1 | - | 11 | 2326 | 96 | ||
| AK091982.1 | - | 1 | 2097 | 96 | ||
| AK122704.1 | - | 10 | 3913 | 96 | ||
| AV661825.1 | - | 417 | 96 | |||
| AY165122.1 | - | 42 | 6662 | 96 | ||
| AY203926.1 | - | 20 | 3727 | 96 | ||
| BC000676.2 | - | 12 | 2080 | 96 | ||
| BC004396.1 | - | 5 | 1084 | 96 | ||
| BC007877.2 | - | 5 | 1400 | 96 | ||
| CR936653.1 | - | 12 | 2614 | 96 | ||
More Information
Set Membership: |
|

MYH14
Genomic Map