Assay Details
| Gene Symbol: ZNF691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017001401.1 | XP_016856890.1 | 1 | 263 | 94 | |
| XM_017001403.1 | XP_016856892.1 | 1 | 262 | 94 | ||
| GenBank mRNA | AK095184.1 | - | 1 | 321 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017001401.1, XM_017001403.1 transcripts.
Set Membership: |
|

ZNF691
Genomic Map