Assay Details
| Gene Symbol: ERBB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005246376.2 | XP_005246433.1 | 1 | 114 | 98 | |
| XM_005246377.2 | XP_005246434.1 | 1 | 114 | 98 | ||
| XM_006712364.2 | XP_006712427.1 | 1 | 114 | 98 | ||
| XM_017003577.1 | XP_016859066.1 | 1 | 114 | 98 | ||
| XM_017003578.1 | XP_016859067.1 | 1 | 114 | 98 | ||
| XM_017003579.1 | XP_016859068.1 | 1 | 114 | 98 | ||
| XM_017003580.1 | XP_016859069.1 | 1 | 114 | 98 | ||
| XM_017003581.1 | XP_016859070.1 | 1 | 114 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005246376.2, XM_005246377.2, XM_006712364.2, XM_017003577.1, XM_017003578.1, XM_017003579.1, XM_017003580.1, XM_017003581.1 transcripts.
Set Membership: |
|

ERBB4
Genomic Map