Assay Details
| Gene Symbol: SCG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003469.4 | NP_003460.2 | 2 | 884 | 195 | |
| GenBank mRNA | AK312452.1 | - | 2 | 745 | 195 | |
| BC022509.1 | - | 2 | 792 | 4820104 | 195 | |
| M25756.1 | - | 2 | 717 | 195 | ||
More Information
Set Membership: |
|

SCG2
Genomic Map