Assay Details
| Gene Symbol: LINC01105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026832.1 | 2 | 9276 | 91 | ||
| GenBank mRNA | AK124750.1 | - | 1 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01105| Gene Symbol: LINC01105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026832.1 | 2 | 9276 | 91 | ||
| GenBank mRNA | AK124750.1 | - | 1 | 91 | ||
Set Membership: |
|