Assay Details
| Gene Symbol: TSSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024248.1 | 1 | 66 | 69 | ||
| GenBank mRNA | AK123648.1 | - | 1 | 66 | 69 | |
| AL528594.3 | - | 1 | 56 | 69 | ||
| DB226687.1 | - | 1 | 66 | 296 | ||
More Information
Set Membership: |
|

TSSC2
Genomic Map