Assay Details
| Gene Symbol: SEMA4F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271661.1 | NP_001258590.1 | 10 | 2724 | 107 | |
| NM_001271662.1 | NP_001258591.1 | 13 | 3090 | 107 | ||
| NM_004263.4 | NP_004254.2 | 14 | 3189 | 107 | ||
| NR_073398.1 | 12 | 2943 | 107 | |||
| NR_073399.1 | 12 | 2996 | 107 | |||
| XM_017003172.1 | XP_016858661.1 | 13 | 3010 | 107 | ||
| XM_017003173.1 | XP_016858662.1 | 12 | 2716 | 107 | ||
| XM_017003174.1 | XP_016858663.1 | 9 | 2571 | 107 | ||
| GenBank mRNA | AK095587.1 | - | 2 | 1287 | 107 | |
| AL136552.1 | - | 14 | 3151 | 107 | ||
More Information
Set Membership: |
|

SEMA4F
Genomic Map