Assay Details
| Gene Symbol: TMPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032283.2 | NP_001027454.1 | 7 - 8 | 1282 | 73 | |
| NM_001307975.1 | NP_001294904.1 | 6 - 7 | 1162 | 73 | ||
| XM_005269132.3 | XP_005269189.1 | 5 - 6 | 1057 | 73 | ||
| GenBank mRNA | BC053675.1 | - | 7 - 8 | 1228 | 6170639 | 73 |
| U09087.1 | - | 7 - 8 | 1224 | 73 | ||
More Information
Set Membership: |
|

TMPO
Genomic Map