Assay Details
| Gene Symbol: TMPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032283.2 | NP_001027454.1 | 8 - 9 | 1372 | 71 | |
| NM_001032284.2 | NP_001027455.1 | 5 - 6 | 1045 | 71 | ||
| NM_001307975.1 | NP_001294904.1 | 7 - 8 | 1252 | 71 | ||
| XM_005269132.3 | XP_005269189.1 | 6 - 7 | 1147 | 71 | ||
| GenBank mRNA | AB209297.1 | - | 5 - 6 | 982 | 71 | |
| AF070631.1 | - | 5 - 6 | 986 | 71 | ||
| AF113682.1 | - | 1 - 2 | 101 | 71 | ||
| AK299708.1 | - | 3 - 4 | 252 | 71 | ||
| BC053675.1 | - | 8 - 9 | 1318 | 6170639 | 71 | |
| U09087.1 | - | 8 - 9 | 1314 | 71 | ||
| U09088.1 | - | 5 - 6 | 987 | 71 | ||
More Information
Set Membership: |
|

TMPO
Genomic Map