Assay Details
| Gene Symbol: KCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287819.1 | NP_001274748.1 | 17 - 18 | 2118 | 98 | |
| NM_001287820.1 | NP_001274749.1 | 16 - 17 | 1968 | 98 | ||
| NM_198503.3 | NP_940905.2 | 17 - 18 | 2118 | 98 | ||
| XM_006711294.2 | XP_006711357.1 | 17 - 18 | 2592 | 98 | ||
| XM_006711295.2 | XP_006711358.1 | 17 - 18 | 2592 | 98 | ||
| XM_011509482.2 | XP_011507784.1 | 16 - 17 | 2517 | 98 | ||
| XM_011509483.2 | XP_011507785.1 | 17 - 18 | 2592 | 98 | ||
| XM_017001178.1 | XP_016856667.1 | 17 - 18 | 2592 | 98 | ||
| XM_017001179.1 | XP_016856668.1 | 17 - 18 | 2592 | 98 | ||
| XM_017001180.1 | XP_016856669.1 | 17 - 18 | 2592 | 98 | ||
| XM_017001181.1 | XP_016856670.1 | 7 - 8 | 1107 | 98 | ||
| XM_017001182.1 | XP_016856671.1 | 16 - 17 | 2483 | 98 | ||
| XM_017001183.1 | XP_016856672.1 | 4 - 5 | 643 | 98 | ||
| XM_017001184.1 | XP_016856673.1 | 16 - 17 | 2483 | 98 | ||
| XM_017001185.1 | XP_016856674.1 | 16 - 17 | 2517 | 98 | ||
| GenBank mRNA | AK127807.1 | - | 10 - 11 | 1468 | 98 | |
| AK131498.1 | - | 16 - 17 | 2193 | 98 | ||
| AK309729.1 | - | 17 - 18 | 2107 | 98 | ||
| AY359444.1 | - | 17 - 18 | 2056 | 98 | ||
| BC103947.1 | - | 13 - 14 | 1499 | 98 | ||
| BC103948.1 | - | 17 - 18 | 2046 | 40009146 | 98 | |
| BC103949.1 | - | 16 - 17 | 1896 | 40009147 | 98 | |
| BC103950.1 | - | 17 - 18 | 2046 | 40009148 | 98 | |
| BX647852.1 | - | 17 - 18 | 2100 | 98 | ||
| EU251076.1 | - | 17 - 18 | 1996 | 98 | ||
More Information
Set Membership: |
|

KCNT2
Genomic Map