Assay Details
| Gene Symbol: TAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031923.3 | NP_114129.1 | 1 - 2 | 375 | 68 | |
| XM_011519741.1 | XP_011518043.1 | 1 - 2 | 402 | 68 | ||
| GenBank mRNA | AJ292190.1 | - | 1 - 2 | 169 | 68 | |
| AK307521.1 | - | 1 - 2 | 351 | 68 | ||
| AK307657.1 | - | 1 - 2 | 353 | 68 | ||
| AL117661.1 | - | 1 - 2 | 103 | 68 | ||
| BC017320.1 | - | 1 - 2 | 361 | 68 | ||
| BC028077.1 | - | 1 - 2 | 332 | 68 | ||
| BC045106.1 | - | 1 - 2 | 361 | 68 | ||
| BC062352.1 | - | 1 - 2 | 342 | 68 | ||
| BC073884.1 | - | 1 - 2 | 366 | 68 | ||
| BC105593.1 | - | 1 - 2 | 379 | 68 | ||
More Information
Set Membership: |
|

TAF3
Genomic Map