Assay Details
| Gene Symbol: NUTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322038.1 | NP_001308967.1 | 3 - 4 | 465 | 112 | |
| NM_001322039.1 | NP_001308968.1 | 3 - 4 | 554 | 112 | ||
| NM_001322040.1 | NP_001308969.1 | 2 - 3 | 464 | 112 | ||
| NM_001322041.1 | NP_001308970.1 | 2 - 3 | 574 | 112 | ||
| NM_005796.2 | NP_005787.1 | 2 - 3 | 375 | 112 | ||
| GenBank mRNA | AK293739.1 | - | 2 - 3 | 174 | 112 | |
| AK311822.1 | - | 2 - 3 | 194 | 112 | ||
| BC002348.2 | - | 2 - 3 | 186 | 2819267 | 112 | |
| BX394845.2 | - | 2 - 3 | 477 | 112 | ||
| CR456937.1 | - | 1 - 2 | 92 | 112 | ||
| HY123109.1 | - | 2 - 3 | 412 | 112 | ||
| U43939.1 | - | 1 - 2 | 92 | 112 | ||
| X07315.1 | - | 2 - 3 | 191 | 112 | ||
More Information
Set Membership: |
|

NUTF2
Genomic Map