Assay Details
| Gene Symbol: ADAM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288973.1 | NP_001275902.1 | 8 - 9 | 1154 | 106 | |
| NM_001288974.1 | NP_001275903.1 | 8 - 9 | 1154 | 106 | ||
| NM_001288975.1 | NP_001275904.1 | 8 - 9 | 1154 | 106 | ||
| NM_003474.5 | NP_003465.3 | 8 - 9 | 1163 | 106 | ||
| NM_021641.4 | NP_067673.2 | 8 - 9 | 1163 | 106 | ||
| XM_017016705.1 | XP_016872194.1 | 7 - 8 | 1066 | 106 | ||
| GenBank mRNA | AF023476.2 | - | 8 - 9 | 1056 | 106 | |
| AF023477.2 | - | 8 - 9 | 1056 | 106 | ||
| AK127856.1 | - | 8 - 9 | 1059 | 106 | ||
| AK291629.1 | - | 8 - 9 | 973 | 106 | ||
| AY358878.1 | - | 8 - 9 | 1051 | 106 | ||
| BC060804.1 | - | 8 - 9 | 1093 | 30348514 | 106 | |
| KF444157.1 | - | 8 - 9 | 1154 | 106 | ||
More Information
Set Membership: |
|

ADAM12
Genomic Map