Assay Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000239.2 | NP_000230.1 | 4 | 1106 | 136 | |
| GenBank mRNA | AK130127.1 | - | 1 | 473 | 136 | |
| AK130149.1 | - | 4 | 1111 | 136 | ||
| BC004147.2 | - | 4 | 1070 | 2959387 | 136 | |
| J03801.1 | - | 4 | 1077 | 136 | ||
| M19045.1 | - | 4 | 1073 | 136 | ||
More Information
Set Membership: |
|

LYZ
Genomic Map