Assay Details
| Gene Symbol: FRY-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103839.1 | 4 | 691 | 103 | ||
| GenBank mRNA | AF086419.1 | - | 1 | 124 | 103 | |
| BC035084.1 | - | 5 | 696 | 103 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16623870 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08759774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206581 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08759766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206577 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206598 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206586 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206588 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206584 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08759781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08759776 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08759765 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206587 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206578 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08759761 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29206602 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 161116 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 908031 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 152958 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 436209 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 444074 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 254431 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 293111 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 217005 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 315658 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 190212 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 267039 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P19766214 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P19766214 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19766233 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19766228 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P11690869 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02749574 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19766214 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19766214 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR1300157620.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157620.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157620.hg.1 | TC1300008548.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008548.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157626.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157626.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157626.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157626.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157626.hg.1 | TC1300008548.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300157624.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157624.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157624.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157624.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157624.hg.1 | TC1300008548.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300157623.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157623.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157623.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157623.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157623.hg.1 | TC1300008548.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300157625.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157625.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157625.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157625.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157625.hg.1 | TC1300008548.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300157622.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157622.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157622.hg.1 | ENST00000428419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157622.hg.1 | NR_103839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157622.hg.1 | TC1300008548.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1558793_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1558793_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558793_at5 | - | - |

FRY-AS1
Genomic Map