Assay Details
| Gene Symbol: LRCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032773.3 | NP_116162.1 | 13 - 14 | 1642 | 87 | |
| XM_005269362.2 | XP_005269419.1 | 13 - 14 | 2013 | 87 | ||
| XM_005269363.2 | XP_005269420.1 | 13 - 14 | 2013 | 87 | ||
| XM_005269365.2 | XP_005269422.1 | 13 - 14 | 2013 | 87 | ||
| XM_005269367.2 | XP_005269424.1 | 13 - 14 | 2013 | 87 | ||
| XM_006713791.3 | XP_006713854.1 | 13 - 14 | 2013 | 87 | ||
| XM_006713792.3 | XP_006713855.1 | 13 - 14 | 2013 | 87 | ||
| XM_006713793.2 | XP_006713856.1 | 13 - 14 | 2013 | 87 | ||
| XM_011513242.2 | XP_011511544.1 | 13 - 14 | 2013 | 87 | ||
| XM_011513243.2 | XP_011511545.1 | 13 - 14 | 2013 | 87 | ||
| XM_017007350.1 | XP_016862839.1 | 13 - 14 | 2013 | 87 | ||
| GenBank mRNA | AK055819.1 | - | 8 - 9 | 748 | 87 | |
| AK293745.1 | - | 13 - 14 | 1631 | 87 | ||
| AK300466.1 | - | 10 - 11 | 1142 | 87 | ||
| AK303521.1 | - | 13 - 14 | 1520 | 87 | ||
| BC007504.2 | - | 13 - 14 | 1594 | 2960711 | 87 | |
More Information
Set Membership: |
|

LRCH3
Genomic Map