Assay Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017006072.1 | XP_016861561.1 | 6 - 7 | 755 | 82 | |
| XM_017006073.1 | XP_016861562.1 | 4 - 5 | 354 | 82 | ||
| XM_017006074.1 | XP_016861563.1 | 6 - 7 | 2773 | 82 | ||
| XM_017006076.1 | XP_016861565.1 | 3 - 4 | 291 | 82 | ||
| XM_017006077.1 | XP_016861566.1 | 5 - 6 | 1702 | 82 | ||
| XM_017006078.1 | XP_016861567.1 | 3 - 4 | 1519 | 82 | ||
| GenBank mRNA | AJ607399.1 | - | 3 - 4 | 256 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017006072.1, XM_017006073.1, XM_017006074.1, XM_017006076.1, XM_017006077.1, XM_017006078.1 transcripts.
Set Membership: |
|

NAALADL2
Genomic Map