Assay Details
| Gene Symbol: UNC13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011517681.2 | XP_011515983.1 | 2 - 3 | 6605 | 76 | |
| XM_011517682.2 | XP_011515984.1 | 2 - 3 | 6606 | 76 | ||
| XM_011517683.2 | XP_011515985.1 | 2 - 3 | 6601 | 76 | ||
| GenBank mRNA | EF139849.1 | - | 1 - 2 | 128 | 76 | |
More Information
Set Membership: |
|

UNC13B
Genomic Map