Assay Details
| Gene Symbol: ZMYM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011542423.2 | XP_011540725.1 | 1 - 2 | 391 | 79 | |
| XM_011542425.2 | XP_011540727.1 | 1 - 2 | 100 | 79 | ||
| GenBank mRNA | BC127113.1 | - | 1 - 2 | 66 | 40125110 | 79 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC127113.1, XM_011542423.2, XM_011542425.2 transcripts.
Set Membership: |
|

ZMYM4
Genomic Map