Assay Details
| Gene Symbol: LINC00083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017003086.1 | XP_016858575.1 | 1 - 2 | 86 | 64 | |
| GenBank mRNA | AK125993.1 | - | 1 - 2 | 97 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK125993.1, XM_017003086.1 transcripts.
Set Membership: |
|

LINC00083
Genomic Map