Assay Details
| Gene Symbol: GALNT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017003259.1 | XP_016858748.1 | 1 - 2 | 47 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017003259.1 transcripts.
Set Membership: |
|

GALNT13
Genomic Map