Assay Details
Gene Symbol: C2orf88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005246905.1 | XP_005246962.1 | 1 - 2 | 200 | 92 | |
XM_011511982.1 | XP_011510284.1 | 1 - 2 | 213 | 92 | ||
XM_011511986.2 | XP_011510288.1 | 1 - 2 | 348 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005246905.1, XM_011511982.1, XM_011511986.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
