Assay Details
| Gene Symbol: SPAG16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024532.4 | NP_078808.3 | 13 - 14 | 1631 | 86 | |
| NR_047659.1 | 15 - 16 | 1806 | 86 | |||
| NR_047660.1 | 12 - 13 | 1512 | 86 | |||
| XM_011511814.1 | XP_011510116.1 | 12 - 13 | 1679 | 86 | ||
| XM_011511815.1 | XP_011510117.1 | 13 - 14 | 1582 | 86 | ||
| XM_011511816.2 | XP_011510118.1 | 13 - 14 | 1582 | 86 | ||
| XM_011511817.1 | XP_011510119.1 | 13 - 14 | 1582 | 86 | ||
| XM_011511818.1 | XP_011510120.1 | 13 - 14 | 1582 | 86 | ||
| XM_011511819.2 | XP_011510121.1 | 12 - 13 | 2330 | 86 | ||
| XM_011511820.1 | XP_011510122.1 | 12 - 13 | 1396 | 86 | ||
| XM_011511821.2 | XP_011510123.1 | 12 - 13 | 2456 | 86 | ||
| XM_011511822.1 | XP_011510124.1 | 11 - 12 | 1312 | 86 | ||
| XM_011511823.2 | XP_011510125.1 | 11 - 12 | 2234 | 86 | ||
| XM_011511835.2 | XP_011510137.1 | 11 - 12 | 1564 | 86 | ||
| XM_017004896.1 | XP_016860385.1 | 14 - 15 | 1711 | 86 | ||
| XM_017004897.1 | XP_016860386.1 | 11 - 12 | 4328 | 86 | ||
| XM_017004898.1 | XP_016860387.1 | 8 - 9 | 1060 | 86 | ||
| XM_017004900.1 | XP_016860389.1 | 12 - 13 | 1463 | 86 | ||
| XM_017004901.1 | XP_016860390.1 | 12 - 13 | 1445 | 86 | ||
| XM_017004902.1 | XP_016860391.1 | 6 - 7 | 781 | 86 | ||
| GenBank mRNA | AF310672.1 | - | 13 - 14 | 1568 | 86 | |
| AF490390.1 | - | 13 - 14 | 1561 | 86 | ||
| AK302351.1 | - | 12 - 13 | 1454 | 86 | ||
| BC025379.1 | - | 15 - 16 | 1811 | 4820489 | 86 | |
| CR749477.1 | - | 11 - 12 | 1317 | 86 | ||
More Information
Set Membership: |
|

SPAG16
Genomic Map