Assay Details
Gene Symbol: NUP50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153645.2 | NP_705931.1 | 2 - 3 | 165 | 84 | |
XM_005261312.1 | XP_005261369.1 | 2 - 3 | 389 | 84 | ||
XM_005261314.1 | XP_005261371.1 | 2 - 3 | 389 | 84 | ||
XM_006724104.1 | XP_006724167.1 | 2 - 3 | 199 | 84 | ||
XM_017028537.1 | XP_016884026.1 | 2 - 3 | 329 | 84 | ||
GenBank mRNA | AL704529.1 | - | 2 - 3 | 84 | ||
BC016055.1 | - | 2 - 3 | 308 | 4856954 | 84 | |
BC028125.1 | - | 2 - 3 | 165 | 5166132 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC016055.1, BC028125.1, NM_153645.2, XM_005261312.1, XM_005261314.1, XM_006724104.1, XM_017028537.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
