Assay Details
| Gene Symbol: LINC00691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026834.1 | 2 | 1644 | 95 | ||
| GenBank mRNA | BC033355.1 | - | 2 | 1635 | 95 | |
| BC039347.1 | - | 2 | 1110 | 95 | ||
| BC045572.1 | - | 2 | 1620 | 95 | ||
More Information
Set Membership: |
|

LINC00691
Genomic Map