| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
136052 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
101113 |
XM_208185.6 |
refseq_mrna_human_XM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
101113 |
XM_940333.2 |
refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
218459 |
AK128104.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
218459 |
BC042592.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
218459 |
BX648119.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
218459 |
NM_178527.2 |
refseq_mrna_human_NM |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3739870 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
45999 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3739870 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P37482527 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P37482527 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P37482527 |
- |
- |