Assay Details
| Gene Symbol: XIAP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BM675621.1 | - | 1 | 207 | 72 | |
| BM931494.1 | - | 2 | 72 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

XIAP-AS1| Gene Symbol: XIAP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BM675621.1 | - | 1 | 207 | 72 | |
| BM931494.1 | - | 2 | 72 | |||
Set Membership: |
|