Assay Details
| Gene Symbol: KCNJ15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276436.1 | NP_001263365.1 | 3 | 290 | 83 | |
| XM_006724002.2 | XP_006724065.1 | 2 | 1867 | 83 | ||
| XM_011529560.2 | XP_011527862.1 | 3 | 313 | 83 | ||
| XM_017028343.1 | XP_016883832.1 | 2 | 442 | 83 | ||
| GenBank mRNA | DB008463.1 | - | 2 | 442 | 83 | |
| DB232867.1 | - | 3 | 264 | 83 | ||
| Y13896.1 | - | 2 | 121 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001276436.1, XM_006724002.2, XM_011529560.2, XM_017028343.1 transcripts.
Set Membership: |
|

KCNJ15
Genomic Map