Assay Details
| Gene Symbol: RNASEH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038429.1 | 1 | 509 | 107 | ||
| NR_038430.1 | 1 | 508 | 107 | |||
| NR_038431.1 | 1 | 508 | 107 | |||
| GenBank mRNA | AK023018.1 | - | 1 | 347 | 107 | |
| BC040057.1 | - | 1 | 403 | 107 | ||
More Information
Set Membership: |
|

RNASEH1-AS1
Genomic Map